Learn More
MSC PRO-SAFE Anti-Fatigue Mat, 16 ft, 3 ft, 1 ea
Catalog No. 5000768679
Encompass
This anti-fatigue mat is designed for dry environments, providing comfort and support for heavy-duty traffic areas. Constructed from a vinyl composite with a vinyl sponge base, it features a diamond plate surface and beveled edges.
  • Measures 16 ft long by 3 ft wide
  • Features 9/16 in thickness
  • Designed for heavy-duty traffic
  • Equipped with beveled edges for enhanced safety
  • Diamond plate surface pattern
  • Non-flammable, complying with FMVSS 302 standards
  • Made from recycled content
  • Suitable for dry area applications
Catalog No. 50-007-68679 Supplier MSC Supplier No. 59747139
This anti-fatigue mat is designed for dry environments, providing comfort and support for heavy-duty traffic areas. Constructed from a vinyl composite with a vinyl sponge base, it features a diamond plate surface and beveled edges.
  • Measures 16 ft long by 3 ft wide
  • Features 9/16 in thickness
  • Designed for heavy-duty traffic
  • Equipped with beveled edges for enhanced safety
  • Diamond plate surface pattern
  • Non-flammable, complying with FMVSS 302 standards
  • Made from recycled content
  • Suitable for dry area applications