Learn More
MSC PRO-SAFE Anti-Fatigue Mat, 3 ft x 2 ft, 1 ea
Catalog No. 5000794939
Encompass
This anti-fatigue mat is designed for heavy-duty use in both dry and wet environments. It features a durable CFR rubber construction and provides comfort and drainage in demanding industrial settings.
  • Dimensions: 3 ft L x 2 ft W
  • Thickness: 7/8 in
  • Constructed from CFR rubber with a composite, single-layer design
  • Straight edges for easy placement
  • Raised circle surface provides enhanced traction and comfort
  • Black color
  • Heavy-duty traffic type for demanding environments
  • Suitable for wet and dry areas
  • Equipped with drainage holes
  • Made from recycled content
Catalog No. 50-007-94939 Supplier MSC Supplier No. 40631384
This anti-fatigue mat is designed for heavy-duty use in both dry and wet environments. It features a durable CFR rubber construction and provides comfort and drainage in demanding industrial settings.
  • Dimensions: 3 ft L x 2 ft W
  • Thickness: 7/8 in
  • Constructed from CFR rubber with a composite, single-layer design
  • Straight edges for easy placement
  • Raised circle surface provides enhanced traction and comfort
  • Black color
  • Heavy-duty traffic type for demanding environments
  • Suitable for wet and dry areas
  • Equipped with drainage holes
  • Made from recycled content