Learn More
MSC PRO-SAFE Anti-Fatigue Mat, 4 x 3 ft., Urethane, 1 ea
Catalog No. 5000787553
Encompass
This anti-fatigue mat is designed for use in dry environments, providing comfort and support for medium-duty traffic areas. It features a durable urethane surface and a composite construction.
  • Beveled edges for safety
  • Pebbled surface pattern
  • Black and yellow color scheme
  • Suitable for dry environments
  • Composite construction with urethane surface and vinyl sponge base
Catalog No. 50-007-87553 Supplier MSC Supplier No. 88266556
This anti-fatigue mat is designed for use in dry environments, providing comfort and support for medium-duty traffic areas. It features a durable urethane surface and a composite construction.
  • Beveled edges for safety
  • Pebbled surface pattern
  • Black and yellow color scheme
  • Suitable for dry environments
  • Composite construction with urethane surface and vinyl sponge base