Learn More
MSC PRO-SAFE Anti-Fatigue Mat, Vinyl, 60 ft Long, 1 ea
Catalog No. 5000788326
Encompass
This anti-fatigue mat is engineered to provide comfort and reduce strain in dry, light-duty work environments. It features a durable vinyl construction designed for prolonged standing.
  • Single layer vinyl sponge base and vinyl surface
  • Provides 3/8 in thickness for optimal comfort
  • Features a 60 ft length and 4 ft width
  • Beveled edges reduce trip hazards
  • Ribbed surface pattern for enhanced traction
  • Suitable for light-duty traffic in dry environments
Catalog No. 50-007-88326 Supplier MSC Supplier No. 88266655
This anti-fatigue mat is engineered to provide comfort and reduce strain in dry, light-duty work environments. It features a durable vinyl construction designed for prolonged standing.
  • Single layer vinyl sponge base and vinyl surface
  • Provides 3/8 in thickness for optimal comfort
  • Features a 60 ft length and 4 ft width
  • Beveled edges reduce trip hazards
  • Ribbed surface pattern for enhanced traction
  • Suitable for light-duty traffic in dry environments