Learn More
MSC PRO-SAFE Anti-Fatigue Matting, 7x3 ft., 9/16 in., 1 ea.
Catalog No. 5000686291
Encompass
This PRO-SAFE anti-fatigue mat is designed for heavy-duty use in dry environments. It features a diamond-plate pattern and composite construction with a vinyl surface and vinyl sponge base, providing comfort and durability.
  • Heavy-duty anti-fatigue matting
  • Designed for dry environments
  • Diamond-plate surface pattern
  • Vinyl surface with vinyl sponge base
  • Beveled edges on all 4 sides
  • Non-flammable
  • Meets FMVSS 302 standard
  • Dimensions: 7 ft. long x 3 ft. wide x 9/16 in. thick
Catalog No. 50-006-86291 Supplier MSC Supplier No. 43080399
This PRO-SAFE anti-fatigue mat is designed for heavy-duty use in dry environments. It features a diamond-plate pattern and composite construction with a vinyl surface and vinyl sponge base, providing comfort and durability.
  • Heavy-duty anti-fatigue matting
  • Designed for dry environments
  • Diamond-plate surface pattern
  • Vinyl surface with vinyl sponge base
  • Beveled edges on all 4 sides
  • Non-flammable
  • Meets FMVSS 302 standard
  • Dimensions: 7 ft. long x 3 ft. wide x 9/16 in. thick