Learn More
MSC PRO-SAFE Arc Flash Pants, 32 cal/sq cm, 1 ea
Catalog No. 5000782693
Encompass
These arc flash pants provide robust protection for personnel in hazardous environments. Designed for safety, they feature durable construction and a high level of arc flash protection.
  • Universal size
  • Leggings garment style
  • Constructed from Indura Ultra Soft material
  • Arc flash garment type
  • Inseam length of 14 in
  • Navy blue color
  • Hazardous protection level 3
  • Maximum arc flash protection of 32 cal/sq cm
Catalog No. 50-007-82693 Supplier MSC Supplier No. 91273714
These arc flash pants provide robust protection for personnel in hazardous environments. Designed for safety, they feature durable construction and a high level of arc flash protection.
  • Universal size
  • Leggings garment style
  • Constructed from Indura Ultra Soft material
  • Arc flash garment type
  • Inseam length of 14 in
  • Navy blue color
  • Hazardous protection level 3
  • Maximum arc flash protection of 32 cal/sq cm