Learn More
MSC Value Collection Ergo One Heel Grounder, Rubber, 1 MegOhm, 10/Pk.
Catalog No. 5000688792
Encompass
The Ergo-One heel grounder provides reliable grounding for mobile personnel when used with conductive or dissipative flooring. Constructed with a dual-layer rubber cup, this grounder features a non-marking interior and a conductive rubber exterior to ensure consistent performance.
  • Features a 1.5 in. x 7.5 in. two-layer rubber cup design
  • Includes an 18 in. conductive nylon ribbon with interlaced silver thread
  • Equipped with a 3/4 in. x 7 in. stretch hook and loop fastening system for a comfortable fit
  • Meets ANSI/ESD S20.20 and MIL 263 standards
  • Symmetrical and reversible design
Catalog No. 50-006-88792 Supplier MSC Supplier No. 97587083
The Ergo-One heel grounder provides reliable grounding for mobile personnel when used with conductive or dissipative flooring. Constructed with a dual-layer rubber cup, this grounder features a non-marking interior and a conductive rubber exterior to ensure consistent performance.
  • Features a 1.5 in. x 7.5 in. two-layer rubber cup design
  • Includes an 18 in. conductive nylon ribbon with interlaced silver thread
  • Equipped with a 3/4 in. x 7 in. stretch hook and loop fastening system for a comfortable fit
  • Meets ANSI/ESD S20.20 and MIL 263 standards
  • Symmetrical and reversible design