Learn More
MSC PRO-SAFE Barricade, 'CAUTION', 3 in. x 1000 ft., 1 ea
Catalog No. 5000685503
Encompass
This polypropylene barricade tape displays general caution messaging for restricting access in a wide range of situations where it is not necessary to provide specific information to pedestrians.
  • General purpose use
  • Made from polypropylene
  • Features 4 mil thickness
  • Measures 3 in. width
  • Offers 1000 ft. length
  • Displays 'CAUTION' message
  • Meets OSHA standards
Catalog No. 50-006-85503 Supplier MSC Supplier No. 63333678
This polypropylene barricade tape displays general caution messaging for restricting access in a wide range of situations where it is not necessary to provide specific information to pedestrians.
  • General purpose use
  • Made from polypropylene
  • Features 4 mil thickness
  • Measures 3 in. width
  • Offers 1000 ft. length
  • Displays 'CAUTION' message
  • Meets OSHA standards