Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PRO-SAFE Barricade, Reflective, Plastic, 2 x 8 in., 1 ea

Catalog No. 5000803147
Encompass

This barricade rail is designed for safety applications, offering visibility and durability. It is suitable for directing traffic or marking hazardous areas.

  • Made from durable plastic
  • Reflective for enhanced visibility
  • Dimensions: 2 in. W x 8 in. H
  • Complies with NCHRP-350 and MUTCD standards
  • Lightweight at 3 lbs.
  • Features a white and orange color scheme

Catalog No. 50-008-03147 Supplier MSC Supplier No. 50892454
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This barricade rail is designed for safety applications, offering visibility and durability. It is suitable for directing traffic or marking hazardous areas.

  • Made from durable plastic
  • Reflective for enhanced visibility
  • Dimensions: 2 in. W x 8 in. H
  • Complies with NCHRP-350 and MUTCD standards
  • Lightweight at 3 lbs.
  • Features a white and orange color scheme

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.