Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PRO-SAFE Barricade Tape, Black/Yellow, 'Shipping', 2x150 ft., 1 Roll

Catalog No. 5000674572
Encompass

Barricade tape is a highly visible and durable plastic tape used to mark off hazardous areas, restrict access, or signal potential dangers in a variety of environments. It is often seen in construction zones, emergency sites, or public safety areas.

  • Material: Woven polyethylene
  • Legend: Shipping
  • Color: Black and yellow
  • Roll length: 150 ft
  • Roll width: 2 in
  • Applications: General
  • Language: English

Catalog No. 50-006-74572 Supplier MSC Supplier No. 57349698
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barricade tape is a highly visible and durable plastic tape used to mark off hazardous areas, restrict access, or signal potential dangers in a variety of environments. It is often seen in construction zones, emergency sites, or public safety areas.

  • Material: Woven polyethylene
  • Legend: Shipping
  • Color: Black and yellow
  • Roll length: 150 ft
  • Roll width: 2 in
  • Applications: General
  • Language: English

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.