Learn More
MSC PRO-SAFE Barricade Tape, Danger, 3x1000 ft, 1 ea
Catalog No. 5000792525
Encompass
PRO-SAFE barricade tape is a highly visible and durable plastic tape designed for temporary placement around hazardous areas to restrict access or signal potential dangers. It helps visually delineate areas requiring caution, reducing the risk of accidents or injuries, aligning with OSHA's definition of a barricade.
  • Made from polyethylene
  • Features 'Danger' message
  • Black and red colors
  • Width of 3 in
  • Length of 1000 ft
  • Helps visually delineate areas
  • Reduces risk of accidents or injuries
  • Suitable for construction zones, emergency sites, and public safety areas
Catalog No. 50-007-92525 Supplier MSC Supplier No. 57349839
PRO-SAFE barricade tape is a highly visible and durable plastic tape designed for temporary placement around hazardous areas to restrict access or signal potential dangers. It helps visually delineate areas requiring caution, reducing the risk of accidents or injuries, aligning with OSHA's definition of a barricade.
  • Made from polyethylene
  • Features 'Danger' message
  • Black and red colors
  • Width of 3 in
  • Length of 1000 ft
  • Helps visually delineate areas
  • Reduces risk of accidents or injuries
  • Suitable for construction zones, emergency sites, and public safety areas