Learn More
MSC PRO-SAFE Bench Grinder Shield, Polycarbonate, 1 ea
Catalog No. 5000766400
Encompass
This bench grinder safety shield features an oversized viewing window and a versatile gooseneck arm, allowing precise positioning. Its large polycarbonate viewing window offers over 58 square inches of protection.
  • Oversized viewing window
  • Adjustable gooseneck arm for customized positioning
  • Budget-friendly and easy to replace
  • Comes fully assembled with two 1/4-20 screws for mounting
  • Bolts directly onto the wheel cover
Catalog No. 50-007-66400 Supplier MSC Supplier No. 83968248
This bench grinder safety shield features an oversized viewing window and a versatile gooseneck arm, allowing precise positioning. Its large polycarbonate viewing window offers over 58 square inches of protection.
  • Oversized viewing window
  • Adjustable gooseneck arm for customized positioning
  • Budget-friendly and easy to replace
  • Comes fully assembled with two 1/4-20 screws for mounting
  • Bolts directly onto the wheel cover