Learn More
MSC PRO-SAFE Bouffant Cap, Polypropylene, White, 21 in, 1000 Pack
Catalog No. 5000793284
Encompass
These disposable bouffant caps provide reliable head covering for use in controlled environments. Constructed from lightweight polypropylene, they offer comfortable wear and effective containment.
  • Lightweight polypropylene construction
  • Latex free
  • ISO Class 6 compatible
  • White color
  • Packaged as 100 per bag, 10 bags per case
  • Size 21 in
Catalog No. 50-007-93284 Supplier MSC Supplier No. 73589830
These disposable bouffant caps provide reliable head covering for use in controlled environments. Constructed from lightweight polypropylene, they offer comfortable wear and effective containment.
  • Lightweight polypropylene construction
  • Latex free
  • ISO Class 6 compatible
  • White color
  • Packaged as 100 per bag, 10 bags per case
  • Size 21 in