Learn More
MSC PRO-SAFE Bouffant, Polypropylene, Latex Free, 21 in, 1000 Count
Catalog No. 5000794701
Encompass
PRO-SAFE bouffant caps are designed for general-purpose use in environments requiring hair containment. They are made from pleated polypropylene and are 100% latex-free, offering comfort and reliability.
  • Made from pleated polypropylene for breathability
  • Fully latex-free to prevent allergic reactions
  • Provides 21 inch size for ample coverage
  • Supplied as 1000 per case for bulk needs
Catalog No. 50-007-94701 Supplier MSC Supplier No. 63582142
PRO-SAFE bouffant caps are designed for general-purpose use in environments requiring hair containment. They are made from pleated polypropylene and are 100% latex-free, offering comfort and reliability.
  • Made from pleated polypropylene for breathability
  • Fully latex-free to prevent allergic reactions
  • Provides 21 inch size for ample coverage
  • Supplied as 1000 per case for bulk needs