Learn More
MSC PRO-SAFE Bumper Guard, 39 in, Black and Yellow, 1 ea
Catalog No. 5000793122
Encompass
Edge Guards protect heads and elbows as well as products and vehicles. This edge profile can be easily clipped on to sharp edges, reducing the risk of injuries and cuts. It is suitable for grooves as small as 1 to 2 mm and can easily be detached and re-used. This edge guard is held in place by the natural pressure created by the guard.
  • Edge guards protect heads, elbows, products, and vehicles.
  • Easily clips onto sharp edges, reducing injury risk.
  • Suitable for grooves as small as 1 to 2 mm.
  • Can be easily detached and re-used.
  • Held in place by natural pressure.
Catalog No. 50-007-93122 Supplier MSC Supplier No. 58406083
Edge Guards protect heads and elbows as well as products and vehicles. This edge profile can be easily clipped on to sharp edges, reducing the risk of injuries and cuts. It is suitable for grooves as small as 1 to 2 mm and can easily be detached and re-used. This edge guard is held in place by the natural pressure created by the guard.
  • Edge guards protect heads, elbows, products, and vehicles.
  • Easily clips onto sharp edges, reducing injury risk.
  • Suitable for grooves as small as 1 to 2 mm.
  • Can be easily detached and re-used.
  • Held in place by natural pressure.