Learn More
MSC PRO-SAFE Chemical Gloves, Polyvinylchloride, Medium, Supported, 12.00 in, 1 ea
Catalog No. 5000802460
Encompass
PRO-SAFE chemical resistant gloves are designed for hand protection in environments requiring chemical resistance. These blue, medium-sized gloves are constructed from polyvinylchloride with a textured finish and cotton lining, offering supported strength.
  • Made from polyvinylchloride material
  • Textured finish for enhanced grip
  • Cotton lining for comfort and support
  • Supported design for additional strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Safety cuff style
  • ANSI/ISEA Abrasion Resistance Level: 4
  • ANSI/ISEA Cut Resistance Level: 0
  • ANSI/ISEA Puncture Resistance Level: 1
  • Lined and seamless construction
  • Overall length: 12.00 in
Catalog No. 50-008-02460 Supplier MSC Supplier No. 01326420
PRO-SAFE chemical resistant gloves are designed for hand protection in environments requiring chemical resistance. These blue, medium-sized gloves are constructed from polyvinylchloride with a textured finish and cotton lining, offering supported strength.
  • Made from polyvinylchloride material
  • Textured finish for enhanced grip
  • Cotton lining for comfort and support
  • Supported design for additional strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Safety cuff style
  • ANSI/ISEA Abrasion Resistance Level: 4
  • ANSI/ISEA Cut Resistance Level: 0
  • ANSI/ISEA Puncture Resistance Level: 1
  • Lined and seamless construction
  • Overall length: 12.00 in