Learn More
MSC PRO-SAFE Chemical Resistant Gloves, Nitrile, 22 mil, Large, 1 Pair
Catalog No. 5000796235
Encompass
PRO-SAFE chemical resistant gloves are designed for tasks requiring dexterity in light-to-medium chemical applications. These green, unlined nitrile gloves feature a raised diamond grip and are FDA approved.
  • Nitrile material
  • Size large
  • Thickness of 22 mil
  • Unsupported design
  • Raised diamond grip surface
  • Straight cuff style
  • ANSI abrasion resistance level 4
  • ANSI cut resistance level 0
  • ANSI puncture resistance level 2
  • FDA approved
  • Overall length of 17.00 in
Catalog No. 50-007-96235 Supplier MSC Supplier No. 02482925
PRO-SAFE chemical resistant gloves are designed for tasks requiring dexterity in light-to-medium chemical applications. These green, unlined nitrile gloves feature a raised diamond grip and are FDA approved.
  • Nitrile material
  • Size large
  • Thickness of 22 mil
  • Unsupported design
  • Raised diamond grip surface
  • Straight cuff style
  • ANSI abrasion resistance level 4
  • ANSI cut resistance level 0
  • ANSI puncture resistance level 2
  • FDA approved
  • Overall length of 17.00 in