Learn More
MSC PRO-SAFE Clean Room Mat, Tacky, 26x45 in., 1 ea
Catalog No. 5000687521
Encompass
This clean room mat features tacky sheets designed to capture contaminants from footwear and cart wheels. It is suitable for dry environments, helping to maintain cleanliness in controlled spaces.
  • Tacky sheets surface material
  • Contains 30 layers per mat
  • Blue color
  • Smooth surface pattern
  • Not reusable
  • No adhesive backing
  • For dry environments
Catalog No. 50-006-87521 Supplier MSC Supplier No. 53136941
This clean room mat features tacky sheets designed to capture contaminants from footwear and cart wheels. It is suitable for dry environments, helping to maintain cleanliness in controlled spaces.
  • Tacky sheets surface material
  • Contains 30 layers per mat
  • Blue color
  • Smooth surface pattern
  • Not reusable
  • No adhesive backing
  • For dry environments