Learn More
MSC PRO-SAFE Coat, Medium, Aluminized Kevlar, Snaps Closure, 1 ea
Catalog No. 5000767283
Encompass
This PRO-SAFE coat provides protection, featuring a medium size to fit a 40 to 42 in. chest. It is constructed from Aluminized Kevlar material.
  • Medium size
  • Chest size: 40 to 42 in.
  • Aluminized Kevlar material
  • Material weight: 19 oz
  • Snaps closure
  • Silver and yellow color
  • Non-hazardous protection
  • Made in the United States
Catalog No. 50-007-67283 Supplier MSC Supplier No. 54164454
This PRO-SAFE coat provides protection, featuring a medium size to fit a 40 to 42 in. chest. It is constructed from Aluminized Kevlar material.
  • Medium size
  • Chest size: 40 to 42 in.
  • Aluminized Kevlar material
  • Material weight: 19 oz
  • Snaps closure
  • Silver and yellow color
  • Non-hazardous protection
  • Made in the United States