Learn More
MSC PRO-SAFE Coat, Non-Hazardous Protection, 2X-Large, Kevlar, 1 ea
Catalog No. 5000789108
Encompass
This PRO-SAFE coat is designed for non-hazardous protection, offering flame-resistant and aluminized properties. It is constructed from Kevlar material, suitable for demanding environments requiring protective outerwear.
  • Made from Kevlar material
  • Provides non-hazardous protection
  • Flame-resistant, flame-retardant, and aluminized garment type
  • Features snaps closure
  • Chest size range of 52 to 54 in.
Catalog No. 50-007-89108 Supplier MSC Supplier No. 56073752
This PRO-SAFE coat is designed for non-hazardous protection, offering flame-resistant and aluminized properties. It is constructed from Kevlar material, suitable for demanding environments requiring protective outerwear.
  • Made from Kevlar material
  • Provides non-hazardous protection
  • Flame-resistant, flame-retardant, and aluminized garment type
  • Features snaps closure
  • Chest size range of 52 to 54 in.