Learn More
MSC PRO-SAFE Coat, X-Large, Aluminized Kevlar, Snaps Closure, 1 Each
Catalog No. 5000788867
Encompass
This PRO-SAFE coat provides personal protection, offering non-hazardous defense and resistance to 2000 deg F radiant heat. It is constructed from aluminized Kevlar with a convenient snaps closure.
  • X-large size
  • Fits 48 to 50 inch chest
  • Constructed from aluminized Kevlar
  • Features snaps closure
  • Provides non-hazardous protection
  • Resistant to 2000 deg F radiant heat
  • Material weight of 19 oz
  • Color: Silver and yellow
Catalog No. 50-007-88867 Supplier MSC Supplier No. 54164512
This PRO-SAFE coat provides personal protection, offering non-hazardous defense and resistance to 2000 deg F radiant heat. It is constructed from aluminized Kevlar with a convenient snaps closure.
  • X-large size
  • Fits 48 to 50 inch chest
  • Constructed from aluminized Kevlar
  • Features snaps closure
  • Provides non-hazardous protection
  • Resistant to 2000 deg F radiant heat
  • Material weight of 19 oz
  • Color: Silver and yellow