Learn More
MSC PRO-SAFE D-Ring Anchor Kit, Alloy Steel, 5000 lb, 1 ea
Catalog No. 5000766900
Encompass
This anchorage connector, commonly referred to as a tie-off point, is designed to join a connecting device to an anchorage when a direct connection does not exist. It is suitable for structural members such as I-beams or columns, and is engineered to support at least 5,000 lbs of force per worker for fall protection applications.
  • Drop forged alloy steel D-rings provide an OSHA-compliant, snaphook-compatible anchor point
  • Supports at least 5,000 lbs when bolted to a structural support
  • Nut and bolt included for installation
Catalog No. 50-007-66900 Supplier MSC Supplier No. 62846944
This anchorage connector, commonly referred to as a tie-off point, is designed to join a connecting device to an anchorage when a direct connection does not exist. It is suitable for structural members such as I-beams or columns, and is engineered to support at least 5,000 lbs of force per worker for fall protection applications.
  • Drop forged alloy steel D-rings provide an OSHA-compliant, snaphook-compatible anchor point
  • Supports at least 5,000 lbs when bolted to a structural support
  • Nut and bolt included for installation