Learn More
MSC PRO-SAFE Decontamination Shower, Inflatable, 2 Lanes, PVC, 1 ea
Catalog No. 5000673165
Encompass
The PRO-SAFE inflatable decontamination shower provides a rapid deployment solution for emergency response and hazardous material decontamination. This portable unit is designed with two lanes to facilitate efficient personnel processing.
  • Inflatable design allows for quick setup and takedown
  • Constructed from durable PVC material for longevity
  • Features two separate lanes for streamlined decontamination procedures
  • Includes an electric inflator/deflator, repair kit, and SCBA inlet hose for comprehensive functionality
Catalog No. 50-006-73165 Supplier MSC Supplier No. 31236284
The PRO-SAFE inflatable decontamination shower provides a rapid deployment solution for emergency response and hazardous material decontamination. This portable unit is designed with two lanes to facilitate efficient personnel processing.
  • Inflatable design allows for quick setup and takedown
  • Constructed from durable PVC material for longevity
  • Features two separate lanes for streamlined decontamination procedures
  • Includes an electric inflator/deflator, repair kit, and SCBA inlet hose for comprehensive functionality