Learn More
MSC PRO-SAFE Disposable Coveralls, Chemical Protection, 5X-Large, Film Laminate, 25/Pk.
Catalog No. 5000781242
Encompass
These disposable coveralls provide chemical protection for various industrial and laboratory environments. Constructed from film laminate material, they offer defense against chemical splash and particulate matter. Designed for comfort and durability, these coveralls feature elasticized cuffs and ankles for a secure fit, and a convenient zipper closure.
  • Designed for chemical protection
  • Made from durable film laminate material
  • Features a zipper closure for easy donning and doffing
  • Serged seams provide enhanced durability
  • Elastic cuffs and ankles ensure a secure and comfortable fit
  • Rated for ISO Class 5 environments
  • Meets ANSI/ISEA 101-1996 standards
Catalog No. 50-007-81242 Supplier MSC Supplier No. 30104293
These disposable coveralls provide chemical protection for various industrial and laboratory environments. Constructed from film laminate material, they offer defense against chemical splash and particulate matter. Designed for comfort and durability, these coveralls feature elasticized cuffs and ankles for a secure fit, and a convenient zipper closure.
  • Designed for chemical protection
  • Made from durable film laminate material
  • Features a zipper closure for easy donning and doffing
  • Serged seams provide enhanced durability
  • Elastic cuffs and ankles ensure a secure and comfortable fit
  • Rated for ISO Class 5 environments
  • Meets ANSI/ISEA 101-1996 standards