Learn More
MSC PRO-SAFE Disposable Coveralls, Chemical Protection, 5X-Large Size, Film Laminate, 25/Pk.
Catalog No. 5000686360
Encompass
This disposable coverall provides chemical protection for various industrial and laboratory applications. It is designed for single-use scenarios, ensuring safety and compliance.
  • Designed for chemical protection
  • Chemical resistant garment type
  • Coverall garment style
  • Features a 5X-Large size for extensive coverage
  • Constructed from durable film laminate material
  • Features a secure zipper closure
  • Elastic cuff style for a snug fit
  • Open ankle style for ease of movement
  • Serged seam style for added durability
  • Meets ISO Class 5 standards
  • Lightweight with a material weight of 1.6000 oz
Catalog No. 50-006-86360 Supplier MSC Supplier No. 73706855
This disposable coverall provides chemical protection for various industrial and laboratory applications. It is designed for single-use scenarios, ensuring safety and compliance.
  • Designed for chemical protection
  • Chemical resistant garment type
  • Coverall garment style
  • Features a 5X-Large size for extensive coverage
  • Constructed from durable film laminate material
  • Features a secure zipper closure
  • Elastic cuff style for a snug fit
  • Open ankle style for ease of movement
  • Serged seam style for added durability
  • Meets ISO Class 5 standards
  • Lightweight with a material weight of 1.6000 oz