Learn More
MSC PRO-SAFE Disposable Coveralls, Chemical Protection, X-Large, Film Laminate, 25/Pk
Catalog No. 5000782804
Encompass
These disposable coveralls are designed for chemical protection, offering reliable safety in demanding environments. They feature a durable film laminate material and are rated for ISO Class 5.
  • X-Large size
  • Zipper closure
  • Serged seam
  • Elastic cuffs
  • Elastic ankles
  • Bunny suit design
  • Material weight 1.87 oz
  • Conforms to ANSI/ISEA 101-1996 standard
Catalog No. 50-007-82804 Supplier MSC Supplier No. 82799263
These disposable coveralls are designed for chemical protection, offering reliable safety in demanding environments. They feature a durable film laminate material and are rated for ISO Class 5.
  • X-Large size
  • Zipper closure
  • Serged seam
  • Elastic cuffs
  • Elastic ankles
  • Bunny suit design
  • Material weight 1.87 oz
  • Conforms to ANSI/ISEA 101-1996 standard