Learn More
MSC PRO-SAFE Disposable Sleeve, Microporous Film, Universal Size, White, 100 Pairs
Catalog No. 5000782231
Encompass
These disposable sleeves, constructed from microporous film, offer protection for arms in various laboratory and industrial settings. They are designed for a universal fit and feature a clean white color.
  • Microporous film material for barrier protection
  • Universal size for broad applicability
  • White color for easy visibility of contaminants
  • Elastic openings at both ends for a secure fit
  • Overall length of 18.0000 in
  • Overall width of 9.0000 in
  • Supplied as 100 pairs
Catalog No. 50-007-82231 Supplier MSC Supplier No. 82799214
These disposable sleeves, constructed from microporous film, offer protection for arms in various laboratory and industrial settings. They are designed for a universal fit and feature a clean white color.
  • Microporous film material for barrier protection
  • Universal size for broad applicability
  • White color for easy visibility of contaminants
  • Elastic openings at both ends for a secure fit
  • Overall length of 18.0000 in
  • Overall width of 9.0000 in
  • Supplied as 100 pairs