Learn More
MSC PRO-SAFE Drum Liner, 55 gal., 18 mil, LDPE, Anti-static, 15/Pack
Catalog No. 5000796416
Encompass
This product consists of rigid, smooth drum liners designed for 55-gallon drums, made from 18 mil low-density polyethylene (LDPE) and featuring anti-static properties for safe handling.
  • Smooth, rigid construction
  • Provides 18 mil thickness for durability
  • Constructed from low-density polyethylene (LDPE)
  • Features anti-static properties
  • Designed for 55-gallon drums
Catalog No. 50-007-96416 Supplier MSC Supplier No. 07509771
This product consists of rigid, smooth drum liners designed for 55-gallon drums, made from 18 mil low-density polyethylene (LDPE) and featuring anti-static properties for safe handling.
  • Smooth, rigid construction
  • Provides 18 mil thickness for durability
  • Constructed from low-density polyethylene (LDPE)
  • Features anti-static properties
  • Designed for 55-gallon drums