Learn More
MSC PRO-SAFE Emergency Shower, 20 GPM, ANSI Z358.1-2004, 1 ea
Catalog No. 5000766276
Encompass
This drench shower and eyewash station provides essential emergency safety equipment. Designed for high-speed applications, it offers immediate decontamination in hazardous environments.
  • Features a plastic bowl and triangular pull rod
  • Push flag activation for quick operation
  • Constructed with galvanized steel pipe
  • Equipped with a plastic shower head
  • Corrosion resistant design
  • Supports top or mid water supply
Catalog No. 50-007-66276 Supplier MSC Supplier No. 86205481
This drench shower and eyewash station provides essential emergency safety equipment. Designed for high-speed applications, it offers immediate decontamination in hazardous environments.
  • Features a plastic bowl and triangular pull rod
  • Push flag activation for quick operation
  • Constructed with galvanized steel pipe
  • Equipped with a plastic shower head
  • Corrosion resistant design
  • Supports top or mid water supply