Learn More
MSC PRO-SAFE ESD Constant Monitor, Continuous Monitoring, AC/Battery Powered, 1 ea
Catalog No. 5000674773
Encompass
The PRO-SAFE PS-1700CM is an ESD Constant Monitor designed to continuously monitor a user's path to ground and the single-wire wrist strap worn by the operator. If the user's ground path is broken, the device emits a loud buzzer and displays a blinking red LED light. Even when switched off, the operator remains connected to earth ground, making the equipment a common point ground.
  • Provides 100% continuous monitoring
  • Tests one person and one wrist strap
  • Red and green LED and fail buzzer
  • Works with single wire capacitance wrist straps
  • Can be calibrated
  • Powered by 110V AC adapter or 9V battery
  • Ideal for in-plant or field service work
  • Ground dock included
  • Made in the USA
  • NIST calibration included
  • CE mark
  • Conforms to ESD S.20.20 spec standards
Catalog No. 50-006-74773 Supplier MSC Supplier No. 57978041
The PRO-SAFE PS-1700CM is an ESD Constant Monitor designed to continuously monitor a user's path to ground and the single-wire wrist strap worn by the operator. If the user's ground path is broken, the device emits a loud buzzer and displays a blinking red LED light. Even when switched off, the operator remains connected to earth ground, making the equipment a common point ground.
  • Provides 100% continuous monitoring
  • Tests one person and one wrist strap
  • Red and green LED and fail buzzer
  • Works with single wire capacitance wrist straps
  • Can be calibrated
  • Powered by 110V AC adapter or 9V battery
  • Ideal for in-plant or field service work
  • Ground dock included
  • Made in the USA
  • NIST calibration included
  • CE mark
  • Conforms to ESD S.20.20 spec standards