Learn More
MSC PRO-SAFE Fall Protection Harness, 350 Lb, X-Large, 1 ea
Catalog No. 5000792898
Encompass
This full-body fall protection harness is designed with a 350 lb load capacity. It features back and side D-rings, durable polyester construction, a friction buckle shoulder strap, and pass-through leg straps.
  • Full body design
  • X-large size
  • Back and side D-rings
  • Polyester material
  • Anti-tangle strap
  • Load capacity of 350 lb
  • Pass-through leg strap buckle
  • Friction shoulder strap buckle
Catalog No. 50-007-92898 Supplier MSC Supplier No. 62846845
This full-body fall protection harness is designed with a 350 lb load capacity. It features back and side D-rings, durable polyester construction, a friction buckle shoulder strap, and pass-through leg straps.
  • Full body design
  • X-large size
  • Back and side D-rings
  • Polyester material
  • Anti-tangle strap
  • Load capacity of 350 lb
  • Pass-through leg strap buckle
  • Friction shoulder strap buckle