Learn More
MSC PRO-SAFE First Aid Kit, 164-Pc, 50-Person, ANSI A, 1 ea
Catalog No. 5000765829
Encompass
This first aid kit is designed for up to 50 people and contains 164 essential pieces for workplace injuries. It is portable and complies with ANSI 2021 Class A standards, suitable for addressing major and minor wounds, burns, and eye injuries.
  • Provides essential supplies for common workplace injuries
  • ANSI 2021 Class A compliant for robust first aid readiness
  • Portable plastic case for easy transport and storage
  • Includes bandages, antiseptics, burn care, and pain relief
  • Contains specialized items like eye wash, CPR mask, and cold pack
Catalog No. 50-007-65829 Supplier MSC Supplier No. 96209689
This first aid kit is designed for up to 50 people and contains 164 essential pieces for workplace injuries. It is portable and complies with ANSI 2021 Class A standards, suitable for addressing major and minor wounds, burns, and eye injuries.
  • Provides essential supplies for common workplace injuries
  • ANSI 2021 Class A compliant for robust first aid readiness
  • Portable plastic case for easy transport and storage
  • Includes bandages, antiseptics, burn care, and pain relief
  • Contains specialized items like eye wash, CPR mask, and cold pack