Learn More
MSC PRO-SAFE Fixed Shade Lens, 4-1/4x2 in, Shade 10, 1 ea
Catalog No. 5000794544
Encompass
This fixed shade lens is designed for welding applications, providing reliable eye protection with a consistent shade level. It features a standard rectangular shape for compatibility with most welding helmets.
  • Fixed shade lens
  • Height: 2 in
  • Width: 4-1/4 in
  • Shade 10 for optimal protection
  • Meets ANSI Z87.1 standards
  • Constructed from polycarbonate
Catalog No. 50-007-94544 Supplier MSC Supplier No. 62674429
This fixed shade lens is designed for welding applications, providing reliable eye protection with a consistent shade level. It features a standard rectangular shape for compatibility with most welding helmets.
  • Fixed shade lens
  • Height: 2 in
  • Width: 4-1/4 in
  • Shade 10 for optimal protection
  • Meets ANSI Z87.1 standards
  • Constructed from polycarbonate