Learn More
MSC PRO-SAFE Flagging Tape, Fluorescent Green, Vinyl, 1-3/16 in., 150 ft.
Catalog No. 5000769726
Encompass
PRO-SAFE flagging tape is a durable vinyl tape designed for various marking applications. It features a fluorescent green color for high visibility, making it suitable for outdoor and industrial environments.
  • Material: vinyl
  • Color: fluorescent green
  • Dimensions: 1-3/16 in. width x 150 ft. length
  • Thickness: 4 mil
Catalog No. 50-007-69726 Supplier MSC Supplier No. 57349797
PRO-SAFE flagging tape is a durable vinyl tape designed for various marking applications. It features a fluorescent green color for high visibility, making it suitable for outdoor and industrial environments.
  • Material: vinyl
  • Color: fluorescent green
  • Dimensions: 1-3/16 in. width x 150 ft. length
  • Thickness: 4 mil