Learn More
MSC PRO-SAFE Flammable Storage Cabinet, 30 gal., Self-Closing, Steel, 1 ea
Catalog No. 5000795636
Encompass
This flammable and hazardous storage cabinet is designed for the safe storage of flammable liquids, ensuring compliance with industry standards NFPA 30, FM, and OSHA.
  • Constructed from steel
  • Self-closing doors
  • Floor mount type
  • Double wall cabinet design
  • Adjustable shelves for flexible storage
  • FM approved for safety
  • Yellow finish for high visibility
  • Capacity of 30 gal.
  • Two doors and one shelf
Catalog No. 50-007-95636 Supplier MSC Supplier No. 03439288
This flammable and hazardous storage cabinet is designed for the safe storage of flammable liquids, ensuring compliance with industry standards NFPA 30, FM, and OSHA.
  • Constructed from steel
  • Self-closing doors
  • Floor mount type
  • Double wall cabinet design
  • Adjustable shelves for flexible storage
  • FM approved for safety
  • Yellow finish for high visibility
  • Capacity of 30 gal.
  • Two doors and one shelf