Learn More
MSC PRO-SAFE Flexible Corner Guard, Yellow/Black, 16.4 ft, 1 ea
Catalog No. 5000794599
Encompass
This PRO-SAFE flexible corner guard is designed for protecting corners in both indoor and outdoor environments. Made from polyurethane foam, it is a Type B surface guard featuring a high-visibility yellow and black color scheme.
  • Flexible polyurethane foam construction
  • Type B surface protection
  • Yellow and black for high visibility
  • Suitable for indoor and outdoor applications
  • Overall length of 16.4 ft
Catalog No. 50-007-94599 Supplier MSC Supplier No. 70289152
This PRO-SAFE flexible corner guard is designed for protecting corners in both indoor and outdoor environments. Made from polyurethane foam, it is a Type B surface guard featuring a high-visibility yellow and black color scheme.
  • Flexible polyurethane foam construction
  • Type B surface protection
  • Yellow and black for high visibility
  • Suitable for indoor and outdoor applications
  • Overall length of 16.4 ft