Learn More
MSC PRO-SAFE Floor and Aisle Marking Tape, 2 in, 100/Pack
Catalog No. 5000803241
Encompass
This floor and aisle marking tape is constructed from durable polyvinylchloride, designed for general traffic areas. It features a 2-inch width and 30-mil thickness, providing a non anti-slip surface for clear delineation.
  • Non anti-slip surface
  • White color
  • PVC material
  • Polyvinylchloride adhesive
  • Width: 2 in
  • Thickness: 30 mil
  • Solid pattern
  • Supplied in a 100 pack
Catalog No. 50-008-03241 Supplier MSC Supplier No. 55224919
This floor and aisle marking tape is constructed from durable polyvinylchloride, designed for general traffic areas. It features a 2-inch width and 30-mil thickness, providing a non anti-slip surface for clear delineation.
  • Non anti-slip surface
  • White color
  • PVC material
  • Polyvinylchloride adhesive
  • Width: 2 in
  • Thickness: 30 mil
  • Solid pattern
  • Supplied in a 100 pack