Learn More
MSC PRO-SAFE Floor and Aisle Marking Tape, 4 in., 1 ea
Catalog No. 5000769509
Encompass
PRO-SAFE Floor & Aisle Marking Tape is a durable polyvinylchloride tape designed for clear and effective floor and aisle marking. With a width of 4 in., a length of 100 ft., and a thickness of 30 mil, it is suitable for general traffic areas.
  • General traffic type
  • Non anti-slip surface
  • Orange color
  • PVC material
  • Polyvinylchloride adhesive
  • Solid pattern
Catalog No. 50-007-69509 Supplier MSC Supplier No. 58284670
PRO-SAFE Floor & Aisle Marking Tape is a durable polyvinylchloride tape designed for clear and effective floor and aisle marking. With a width of 4 in., a length of 100 ft., and a thickness of 30 mil, it is suitable for general traffic areas.
  • General traffic type
  • Non anti-slip surface
  • Orange color
  • PVC material
  • Polyvinylchloride adhesive
  • Solid pattern