Learn More
MSC PRO-SAFE Floor Marking Tape, 2 in, Blue, 100 ft
Catalog No. 5000766927
Encompass
This floor and aisle marking tape is designed for general traffic areas, providing clear visual demarcation. Made from durable polyvinylchloride, it features a non anti-slip surface for enhanced safety.
  • General traffic tape type
  • Non anti-slip surface
  • Blue color
  • PVC polyvinylchloride material
  • Two inch width
  • A thickness of 30 mil
  • One hundred foot length
  • Solid pattern
Catalog No. 50-007-66927 Supplier MSC Supplier No. 55224828
This floor and aisle marking tape is designed for general traffic areas, providing clear visual demarcation. Made from durable polyvinylchloride, it features a non anti-slip surface for enhanced safety.
  • General traffic tape type
  • Non anti-slip surface
  • Blue color
  • PVC polyvinylchloride material
  • Two inch width
  • A thickness of 30 mil
  • One hundred foot length
  • Solid pattern