Learn More
MSC PRO-SAFE Floor Marking Tape, PVC, 2 in Wide, Angles, 100/Pk.
Catalog No. 5000793136
Encompass
PRO-SAFE floor and aisle marking tape is designed for general traffic areas, providing clear visual guidance with its durable construction.
  • Made from PVC material
  • Features a non anti-slip surface
  • Measures 2 in wide
  • Has a thickness of 30 mil
  • Comes in a gray solid pattern
  • Die cut into angle shapes
  • Each pack contains 100 pieces
Catalog No. 50-007-93136 Supplier MSC Supplier No. 58278904
PRO-SAFE floor and aisle marking tape is designed for general traffic areas, providing clear visual guidance with its durable construction.
  • Made from PVC material
  • Features a non anti-slip surface
  • Measures 2 in wide
  • Has a thickness of 30 mil
  • Comes in a gray solid pattern
  • Die cut into angle shapes
  • Each pack contains 100 pieces