Learn More
MSC PRO-SAFE Floor Tape, 4 in. by 100 ft., 1 Roll
Catalog No. 5000673714
Encompass
This PRO-SAFE Floor & Aisle Marking Tape is 4 in. wide and 100 ft. long. It has a 50 mil thickness and is made of vinyl, suitable for various marking applications.
  • Heavy-duty traffic type
  • Smooth surface
  • Rubber adhesive material
  • General application
  • Solid pattern
  • Solid line die cut shape
  • Compliant with ANSI Z535, OSHA 1910.145, OSHA 29 CFR 1910.22, and other standards
  • Country of origin: United States
Catalog No. 50-006-73714 Supplier MSC Supplier No. 44008142
This PRO-SAFE Floor & Aisle Marking Tape is 4 in. wide and 100 ft. long. It has a 50 mil thickness and is made of vinyl, suitable for various marking applications.
  • Heavy-duty traffic type
  • Smooth surface
  • Rubber adhesive material
  • General application
  • Solid pattern
  • Solid line die cut shape
  • Compliant with ANSI Z535, OSHA 1910.145, OSHA 29 CFR 1910.22, and other standards
  • Country of origin: United States