Learn More
MSC PRO-SAFE G-Tek KEV Cut-Resistant Gloves, Medium, 1 Pair
Catalog No. 5000802905
Encompass
PRO-SAFE G-Tek KEV cut-resistant gloves are constructed from Kevlar with a latex-coated palm and fingers, offering a crinkle grip. These gloves provide ANSI/ISEA Cut A3, Puncture 4, and Abrasion 2 protection, making them suitable for various demanding applications.
  • Kevlar material construction provides high cut resistance
  • Latex coating on palm and fingers offers crinkle grip
  • Achieves ANSI/ISEA Cut A3, Puncture 4, and Abrasion 2 ratings
  • Seamless knit construction enhances comfort and dexterity
  • Features an elastic knit cuff for a secure fit
  • Capable of conducting electricity
  • Overall length of 9 in.
  • Size medium, numeric size 8
Catalog No. 50-008-02905 Supplier MSC Supplier No. 84491208
PRO-SAFE G-Tek KEV cut-resistant gloves are constructed from Kevlar with a latex-coated palm and fingers, offering a crinkle grip. These gloves provide ANSI/ISEA Cut A3, Puncture 4, and Abrasion 2 protection, making them suitable for various demanding applications.
  • Kevlar material construction provides high cut resistance
  • Latex coating on palm and fingers offers crinkle grip
  • Achieves ANSI/ISEA Cut A3, Puncture 4, and Abrasion 2 ratings
  • Seamless knit construction enhances comfort and dexterity
  • Features an elastic knit cuff for a secure fit
  • Capable of conducting electricity
  • Overall length of 9 in.
  • Size medium, numeric size 8