Learn More
MSC PRO-SAFE Glove Clip, Polyoxymethylene, Dielectric, Lime, 4-3/4 in, 1 Pair
Catalog No. 5000793381
Encompass
These glove clips feature a belt slot, designed to keep gloves readily accessible and prevent loss. Each clip securely holds one pair of gloves.
  • Made from polyoxymethylene
  • Dielectric
  • Equipped with a belt clip
  • Lime color
  • Measures 4-3/4 x 1-3/4 x 3/4 in
Catalog No. 50-007-93381 Supplier MSC Supplier No. 74930157
These glove clips feature a belt slot, designed to keep gloves readily accessible and prevent loss. Each clip securely holds one pair of gloves.
  • Made from polyoxymethylene
  • Dielectric
  • Equipped with a belt clip
  • Lime color
  • Measures 4-3/4 x 1-3/4 x 3/4 in