Learn More
MSC PRO-SAFE Hairnet, Brown, Nylon, 21 in., 1000/Pk.
Catalog No. 5000765828
Encompass
This hairnet is made of durable nylon and designed for various applications. It has a 21 in. size and meets ISO Class 7 standards.
  • Made of nylon
  • Measures 21 in. in size
  • ISO Class 7
  • Manufactured in China
  • Supplied as a 1000 pack
Catalog No. 50-007-65828 Supplier MSC Supplier No. 95109971
This hairnet is made of durable nylon and designed for various applications. It has a 21 in. size and meets ISO Class 7 standards.
  • Made of nylon
  • Measures 21 in. in size
  • ISO Class 7
  • Manufactured in China
  • Supplied as a 1000 pack