Learn More
MSC PRO-SAFE High Visibility Vest, Large, ANSI Class 2, 1 ea
Catalog No. 5000688146
Encompass
The PRO-SAFE High Visibility Vest is a general-purpose safety vest designed for environments requiring ANSI Class 2 compliance. It features an orange mesh material with two silver horizontal stripes, each 2 in wide, ensuring high visibility. The vest secures with a hook and loop closure.
  • General purpose vest
  • ANSI Class 2 compliant
  • Orange mesh material
  • Two 2 in silver horizontal stripes
  • Hook and loop closure
  • No pockets
  • Manufactured in China
  • Adheres to ANSI/ISEA 107-2015 standards
Catalog No. 50-006-88146 Supplier MSC Supplier No. 87041034
The PRO-SAFE High Visibility Vest is a general-purpose safety vest designed for environments requiring ANSI Class 2 compliance. It features an orange mesh material with two silver horizontal stripes, each 2 in wide, ensuring high visibility. The vest secures with a hook and loop closure.
  • General purpose vest
  • ANSI Class 2 compliant
  • Orange mesh material
  • Two 2 in silver horizontal stripes
  • Hook and loop closure
  • No pockets
  • Manufactured in China
  • Adheres to ANSI/ISEA 107-2015 standards