Learn More
MSC PRO-SAFE High Visibility Vest, Large, ANSI Class 2, 1 ea
Catalog No. 5000675583
Encompass
This high visibility surveyor's vest is designed for safety and functionality, meeting ANSI Class 2 standards. It features a durable, solid material composition and a secure zipper closure, making it ideal for various work environments.
  • Includes 12 pockets
  • ANSI Class 2 compliant
  • Zipper closure type
  • Solid material composition
  • Two horizontal silver stripes, 2 in. wide
  • Approved under ANSI/ISEA 107-2015 standard
Catalog No. 50-006-75583 Supplier MSC Supplier No. 87041406
This high visibility surveyor's vest is designed for safety and functionality, meeting ANSI Class 2 standards. It features a durable, solid material composition and a secure zipper closure, making it ideal for various work environments.
  • Includes 12 pockets
  • ANSI Class 2 compliant
  • Zipper closure type
  • Solid material composition
  • Two horizontal silver stripes, 2 in. wide
  • Approved under ANSI/ISEA 107-2015 standard