Learn More
MSC PRO-SAFE High Visibility Vest, Large, ANSI Class 2, 1 ea
Catalog No. 5000688443
Encompass
This PRO-SAFE high visibility vest is designed for general purpose use, offering enhanced visibility in various work environments. It is a Class 2 vest, ensuring compliance with safety standards.
  • Made from breathable mesh material
  • Features two 2 in. wide horizontal silver stripes for increased visibility
  • Equipped with a hook and loop closure for easy wear
  • Complies with ANSI/ISEA 107-2015 standards
  • Large size for comfortable fit
  • Yellow color for optimal visual recognition
Catalog No. 50-006-88443 Supplier MSC Supplier No. 87041083
This PRO-SAFE high visibility vest is designed for general purpose use, offering enhanced visibility in various work environments. It is a Class 2 vest, ensuring compliance with safety standards.
  • Made from breathable mesh material
  • Features two 2 in. wide horizontal silver stripes for increased visibility
  • Equipped with a hook and loop closure for easy wear
  • Complies with ANSI/ISEA 107-2015 standards
  • Large size for comfortable fit
  • Yellow color for optimal visual recognition