Learn More
MSC PRO-SAFE Jacket, Flame-Resistant Cotton, Medium (38-40 in Chest), 1 ea
Catalog No. 5000795560
Encompass
This PRO-SAFE jacket is a flame-resistant and flame-retardant garment designed for protection in environments requiring specialized safety wear. It features a snaps closure and is suitable for various industrial applications.
  • Flame-resistant and flame-retardant material
  • Snaps closure for secure fastening
  • Medium size for a chest circumference of 38–40 in.
  • Constructed from durable cotton fabric
  • Material weight of 9.00 oz
  • Available in gray and green colors
Catalog No. 50-007-95560 Supplier MSC Supplier No. 42649202
This PRO-SAFE jacket is a flame-resistant and flame-retardant garment designed for protection in environments requiring specialized safety wear. It features a snaps closure and is suitable for various industrial applications.
  • Flame-resistant and flame-retardant material
  • Snaps closure for secure fastening
  • Medium size for a chest circumference of 38–40 in.
  • Constructed from durable cotton fabric
  • Material weight of 9.00 oz
  • Available in gray and green colors