Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PRO-SAFE Jacket, Non-Hazardous Protection, 3X-Large, Cotton, Flame-Resistant, 1 ea

Catalog No. 5000769113
Encompass

This PRO-SAFE jacket provides non-hazardous protection, crafted from cotton and designed for welding applications. It features a snaps closure and is both flame-resistant and flame-retardant.

  • Size: 3X-Large
  • Color: Green
  • Material: Cotton
  • Closure type: Snaps
  • Chest size: 54 to 56 in.
  • Material weight: 9.00 oz
  • Number of pockets: 2
  • Country of origin: Mexico

Catalog No. 50-007-69113 Supplier MSC Supplier No. 82191883
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This PRO-SAFE jacket provides non-hazardous protection, crafted from cotton and designed for welding applications. It features a snaps closure and is both flame-resistant and flame-retardant.

  • Size: 3X-Large
  • Color: Green
  • Material: Cotton
  • Closure type: Snaps
  • Chest size: 54 to 56 in.
  • Material weight: 9.00 oz
  • Number of pockets: 2
  • Country of origin: Mexico

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.