Learn More
MSC PRO-SAFE Lab Coat, 2X-Large, White, Polyolefin, Snap Closure, 30 Pack
Catalog No. 5000792118
Encompass
These PRO-SAFE lab coats are designed for non-cleanroom environments, offering an economical, latex-free solution for various industrial applications. Made from breathable polyolefin, they provide a dependable barrier against dust and light splash.
  • Economical alternative to Tyvek®
  • Latex-free construction
  • Single collar design
  • Snap front closure for secure fit
  • Without pockets for simplified use
  • Breathable polyolefin material
Catalog No. 50-007-92118 Supplier MSC Supplier No. 82799180
These PRO-SAFE lab coats are designed for non-cleanroom environments, offering an economical, latex-free solution for various industrial applications. Made from breathable polyolefin, they provide a dependable barrier against dust and light splash.
  • Economical alternative to Tyvek®
  • Latex-free construction
  • Single collar design
  • Snap front closure for secure fit
  • Without pockets for simplified use
  • Breathable polyolefin material